N-(5-Amino-2-methylphenyl)-2-(3,5-dimethylphenoxy)acetamide

Supplier: Matrix Scientific

025101-500MG
101819-580EA 306.15 USD
101819-580
N-(5-Amino-2-methylphenyl)-2-(3,5-dimethylphenoxy)acetamide
N-(5-Amino-2-methylphenyl)-2-(3,5-dimethylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₂
MDL Number: MFCD09738636
CAS Number: 954251-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR