N-(4-Aminophenyl)propanamide

Supplier: Matrix Scientific

025098-500MG
101819-574EA 70.77 USD
101819-574
N-(4-Aminophenyl)propanamide
N-(4-Aminophenyl)propanamide

Hazard: Irritant

Formula: C₉H₁₂N₂O
MDL Number: MFCD02063079
CAS Number: 59690-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR