N-(3-Aminophenyl)-2-[2,4-di(tert-butyl)phenoxy]acetamide

Supplier: Matrix Scientific

025092-500MG
101819-562EA 306.15 USD
101819-562
N-(3-Aminophenyl)-2-[2,4-di(tert-butyl)phenoxy]acetamide
N-(3-Aminophenyl)-2-[2,4-di(tert-butyl)phenoxy]acetamide

Hazard: Irritant

Formula: C₂₂H₃₀N₂O₂
MW: 354.5 g/mol
MDL Number: MFCD09997111
CAS Number: 1020722-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR