N-(3-Aminophenyl)-3-(isopentyloxy)benzamide

Supplier: Matrix Scientific

025089-500MG
101819-556EA 306.15 USD
101819-556
N-(3-Aminophenyl)-3-(isopentyloxy)benzamide
N-(3-Aminophenyl)-3-(isopentyloxy)benzamide

Hazard: Irritant

Formula: C₁₈H₂₂N₂O₂
MDL Number: MFCD09997108
CAS Number: 1020722-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR