N-(4-Aminophenyl)-2-phenoxyacetamide

Supplier: Matrix Scientific

025087-500MG
101819-552EA 306.15 USD
101819-552
N-(4-Aminophenyl)-2-phenoxyacetamide
N-(4-Aminophenyl)-2-phenoxyacetamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD00446208
CAS Number: 293327-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR