N-(3-Aminophenyl)octanamide

Supplier: Matrix Scientific

025086-500MG
101819-550EA 306.15 USD
101819-550
N-(3-Aminophenyl)octanamide
N-(3-Aminophenyl)octanamide

Hazard: Irritant

Formula: C₁₄H₂₂N₂O
MDL Number: MFCD09997106
CAS Number: 1020722-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR