N-(3-Aminophenyl)-3-butoxybenzamide

Supplier: Matrix Scientific

025083-500MG
101819-544EA 306.15 USD
101819-544
N-(3-Aminophenyl)-3-butoxybenzamide
N-(3-Aminophenyl)-3-butoxybenzamide

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₂
MDL Number: MFCD09997104
CAS Number: 1020722-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR