N-(3-Aminophenyl)-2-(4-isopropylphenoxy)propanamide

Supplier: Matrix Scientific

025081-500MG
101819-540EA 306.15 USD
101819-540
N-(3-Aminophenyl)-2-(4-isopropylphenoxy)propanamide
N-(3-Aminophenyl)-2-(4-isopropylphenoxy)propanamide

Hazard: Irritant

Formula: C₁₈H₂₂N₂O₂
MW: 298.39 g/mol
MDL Number: MFCD09740522
CAS Number: 954576-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR