N-(3-Aminophenyl)-4-(heptyloxy)benzamide

Supplier: Matrix Scientific

025078-500MG
101819-534EA 306.15 USD
101819-534
N-(3-Aminophenyl)-4-(heptyloxy)benzamide
N-(3-Aminophenyl)-4-(heptyloxy)benzamide

Hazard: Irritant

Formula: C₂₀H₂₆N₂O₂
MDL Number: MFCD09997102
CAS Number: 1020722-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR