N-(3-Aminophenyl)-2-fluorobenzamide

Supplier: Matrix Scientific

025077-500MG
101819-532EA 306.15 USD
101819-532
N-(3-Aminophenyl)-2-fluorobenzamide
N-(3-Aminophenyl)-2-fluorobenzamide

Hazard: Irritant

Formula: C₁₃H₁₁FN₂O
MDL Number: MFCD09045999
CAS Number: 926187-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR