N-(3-Aminophenyl)-2-(hexyloxy)benzamide

Supplier: Matrix Scientific

025069-500MG
101819-516EA 306.15 USD
101819-516
N-(3-Aminophenyl)-2-(hexyloxy)benzamide
N-(3-Aminophenyl)-2-(hexyloxy)benzamide

Hazard: Irritant

Formula: C₁₉H₂₄N₂O₂
MDL Number: MFCD09997096
CAS Number: 1020722-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR