N-(3-Aminophenyl)-4-propoxybenzamide

Supplier: Matrix Scientific

025067-500MG
101819-512EA 306.15 USD
101819-512
N-(3-Aminophenyl)-4-propoxybenzamide
N-(3-Aminophenyl)-4-propoxybenzamide

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₂
MDL Number: MFCD09997094
CAS Number: 1020722-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR