N-(3-Aminophenyl)-4-isobutoxybenzamide

Supplier: Matrix Scientific

025066-500MG
101819-510EA 306.15 USD
101819-510
N-(3-Aminophenyl)-4-isobutoxybenzamide
N-(3-Aminophenyl)-4-isobutoxybenzamide

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₂
MDL Number: MFCD09997093
CAS Number: 1020722-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR