N-(3-Aminophenyl)-2-phenoxybutanamide
Supplier: Matrix Scientific
025060-500MG
101819-498EA
306.15
USD
101819-498
N-(3-Aminophenyl)-2-phenoxybutanamide
N-(3-Aminophenyl)-2-phenoxybutanamide
Hazard: Irritant
Formula:
C₁₆H₁₈N₂O₂ |
MDL Number:
MFCD09997090 CAS Number: 1020722-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...