N-(3-Aminophenyl)-2-(3-methylphenoxy)butanamide

Supplier: Matrix Scientific

025059-500MG
101819-496EA 306.15 USD
101819-496
N-(3-Aminophenyl)-2-(3-methylphenoxy)butanamide
N-(3-Aminophenyl)-2-(3-methylphenoxy)butanamide

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₂
MDL Number: MFCD09997089
CAS Number: 1020722-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR