N-(3-Aminophenyl)-2-ethoxybenzamide

Supplier: Matrix Scientific

025058-500MG
101819-494EA 306.15 USD
101819-494
N-(3-Aminophenyl)-2-ethoxybenzamide
N-(3-Aminophenyl)-2-ethoxybenzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD09734748
CAS Number: 953730-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR