N-(3-Aminophenyl)-4-ethoxybenzamide

Supplier: Matrix Scientific

025056-500MG
101819-490EA 306.15 USD
101819-490
N-(3-Aminophenyl)-4-ethoxybenzamide
N-(3-Aminophenyl)-4-ethoxybenzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD09811789
CAS Number: 1016516-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR