N-(3-Aminophenyl)-2-(2-methoxyphenoxy)propanamide

Supplier: Matrix Scientific

025052-500MG
101819-482EA 306.15 USD
101819-482
N-(3-Aminophenyl)-2-(2-methoxyphenoxy)propanamide
N-(3-Aminophenyl)-2-(2-methoxyphenoxy)propanamide

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₃
MDL Number: MFCD09736636
CAS Number: 954252-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR