N-(3-Aminophenyl)-2,2-dimethylpropanamide

Supplier: Matrix Scientific

025041-500MG
101819-460EA 306.15 USD
101819-460
N-(3-Aminophenyl)-2,2-dimethylpropanamide
N-(3-Aminophenyl)-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD09048462
CAS Number: 41402-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR