N-(3-Aminophenyl)-2-(4-methoxyphenoxy)acetamide

Supplier: Matrix Scientific

025039-500MG
101819-456EA 306.15 USD
101819-456
N-(3-Aminophenyl)-2-(4-methoxyphenoxy)acetamide
N-(3-Aminophenyl)-2-(4-methoxyphenoxy)acetamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₃
MDL Number: MFCD09727894
CAS Number: 953719-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR