N-(3-Aminophenyl)-2-(4-ethylphenoxy)acetamide

Supplier: Matrix Scientific

025037-500MG
101819-452EA 306.15 USD
101819-452
N-(3-Aminophenyl)-2-(4-ethylphenoxy)acetamide
N-(3-Aminophenyl)-2-(4-ethylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₂
MDL Number: MFCD09730161
CAS Number: 953723-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR