N-(3-Aminophenyl)-2-(2-isopropyl-5-methylphenoxy)acetamide

Supplier: Matrix Scientific

025034-500MG
101819-446EA 306.15 USD
101819-446
N-(3-Aminophenyl)-2-(2-isopropyl-5-methylphenoxy)acetamide
N-(3-Aminophenyl)-2-(2-isopropyl-5-methylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₈H₂₂N₂O₂
MDL Number: MFCD09730163
CAS Number: 953724-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR