N-(4-Aminophenyl)-2,4-dichlorobenzamide

Supplier: Matrix Scientific

025032-500MG
101819-442EA 306.15 USD
101819-442
N-(4-Aminophenyl)-2,4-dichlorobenzamide
N-(4-Aminophenyl)-2,4-dichlorobenzamide

Hazard: Irritant

Formula: C₁₃H₁₀Cl₂N₂O
MDL Number: MFCD00851342
CAS Number: 293737-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR