N-(3-Aminophenyl)cyclopropanecarboxamide

Supplier: Matrix Scientific

025031-500MG
101819-440EA 461.54 USD
101819-440
N-(3-Aminophenyl)cyclopropanecarboxamide
N-(3-Aminophenyl)cyclopropanecarboxamide

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD08593589
CAS Number: 879127-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR