N-(3-Aminophenyl)-2-(2-methoxyphenoxy)acetamide

Supplier: Matrix Scientific

025030-500MG
101819-438EA 306.15 USD
101819-438
N-(3-Aminophenyl)-2-(2-methoxyphenoxy)acetamide
N-(3-Aminophenyl)-2-(2-methoxyphenoxy)acetamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₃
MDL Number: MFCD09735954
CAS Number: 953896-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR