N-(3-Aminophenyl)-2-(4-methoxyphenyl)acetamide

Supplier: Matrix Scientific

025025-500MG
101819-428EA 306.15 USD
101819-428
N-(3-Aminophenyl)-2-(4-methoxyphenyl)acetamide
N-(3-Aminophenyl)-2-(4-methoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD09809647
CAS Number: 1016766-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR