N-(3-Aminophenyl)-2-(2-methylphenoxy)acetamide

Supplier: Matrix Scientific

025022-500MG
101819-422EA 181.54 USD
101819-422
N-(3-Aminophenyl)-2-(2-methylphenoxy)acetamide
N-(3-Aminophenyl)-2-(2-methylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD00432453
CAS Number: 319489-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR