N-(3-Aminophenyl)-4-methoxybenzamide

Supplier: Matrix Scientific

025017-500MG
101819-412EA 306.15 USD
101819-412
N-(3-Aminophenyl)-4-methoxybenzamide
N-(3-Aminophenyl)-4-methoxybenzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD08691693
CAS Number: 41378-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR