N-(3-Aminophenyl)-2-phenoxyacetamide

Supplier: Matrix Scientific

025014-500MG
101819-406EA 269.23 USD
101819-406
N-(3-Aminophenyl)-2-phenoxyacetamide
N-(3-Aminophenyl)-2-phenoxyacetamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD09465971
CAS Number: 939230-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR