N-(3-Aminophenyl)-3-methylbenzamide

Supplier: Matrix Scientific

025013-500MG
101819-404EA 306.15 USD
101819-404
N-(3-Aminophenyl)-3-methylbenzamide
N-(3-Aminophenyl)-3-methylbenzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD01806272
CAS Number: 585517-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR