N-(3-Aminophenyl)-3-phenylpropanamide

Supplier: Matrix Scientific

025012-500MG
101819-402EA 306.15 USD
101819-402
N-(3-Aminophenyl)-3-phenylpropanamide
N-(3-Aminophenyl)-3-phenylpropanamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD08691856
CAS Number: 754162-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR