N-(3-Aminophenyl)-4-chlorobenzamide

Supplier: Matrix Scientific

025011-500MG
101819-400EA 306.15 USD
101819-400
N-(3-Aminophenyl)-4-chlorobenzamide
N-(3-Aminophenyl)-4-chlorobenzamide

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂O
MDL Number: MFCD01027216
CAS Number: 905811-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR