N-(3-Aminophenyl)-2-chlorobenzamide

Supplier: Matrix Scientific

025009-500MG
101819-396EA 306.15 USD
101819-396
N-(3-Aminophenyl)-2-chlorobenzamide
N-(3-Aminophenyl)-2-chlorobenzamide

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂O
MDL Number: MFCD00297720
CAS Number: 443290-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR