N-(3-Aminophenyl)-2-(2-isopropylphenoxy)acetamide

Supplier: Matrix Scientific

025001-500MG
101819-380EA 306.15 USD
101819-380
N-(3-Aminophenyl)-2-(2-isopropylphenoxy)acetamide
N-(3-Aminophenyl)-2-(2-isopropylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₂
MDL Number: MFCD09742153
CAS Number: 954564-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR