1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]homopiperazine

Supplier: Matrix Scientific

024991-250MG
101819-354EA 361.54 USD
101819-354
1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]homopiperazine
1-[(5-Bromo-2-methyl-4-methylsulfonyl)phenyl]homopiperazine

Hazard: Irritant

Formula: C₁₃H₁₉BrN₂O₂S
MDL Number: MFCD09953243
CAS Number: 1020722-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR