1-[(2-Fluoro-4-phenylsulfonyl)phenyl]homopiperazine

Supplier: Matrix Scientific

024990-250MG
101819-352EA 267.69 USD
101819-352
1-[(2-Fluoro-4-phenylsulfonyl)phenyl]homopiperazine
1-[(2-Fluoro-4-phenylsulfonyl)phenyl]homopiperazine

Hazard: Irritant

Formula: C₁₇H₁₉FN₂O₂S
MDL Number: MFCD09953242
CAS Number: 1020722-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR