1-[(2-Cyano-4-fluoro)phenyl]piperidine-3-carboxylic acid
Supplier: Matrix Scientific
024983-500MG
101819-338EA
278.46
USD
101819-338
1-[(2-Cyano-4-fluoro)phenyl]piperidine-3-carboxylic acid
1-[(2-Cyano-4-fluoro)phenyl]piperidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₃FN₂O₂ MW: 248.26 g/mol |
MDL Number:
MFCD09750917 CAS Number: 1000339-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...