1,3-Dimethyl-1H-pyrazol-5-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 1,3-Dimethyl-1H-pyrazole-5-carboxaldehyde1,3-Dimethyl-1H-pyrazole-5-carbaldehyde

024973-500MG
101819-318EA 186.15 USD
101819-318
1,3-Dimethyl-1H-pyrazol-5-carbaldehyde
1,3-Dimethyl-1H-pyrazol-5-carbaldehyde

Hazard: Irritant

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
MDL Number: MFCD02681907
CAS Number: 25016-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR