1-[(4-Ethylsulfonyl-2-fluoro)phenyl]piperazine

Supplier: Matrix Scientific

024971-250MG
101819-314EA 287.69 USD
101819-314
1-[(4-Ethylsulfonyl-2-fluoro)phenyl]piperazine
1-[(4-Ethylsulfonyl-2-fluoro)phenyl]piperazine

Hazard: Irritant

Formula: C₁₂H₁₇FN₂O₂S
MDL Number: MFCD09038323
CAS Number: 845617-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR