2-tert-Butyl-7-(1-methylpiperazin-4-yl)-4-nitroindole

Supplier: Matrix Scientific

024969-250MG
101819-310EA 467.69 USD
101819-310
2-tert-Butyl-7-(1-methylpiperazin-4-yl)-4-nitroindole
2-tert-Butyl-7-(1-methylpiperazin-4-yl)-4-nitroindole

Hazard: Irritant

Formula: C₁₇H₂₄N₄O₂
MDL Number: MFCD09997082
CAS Number: 1020722-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR