1-(6-Chloropyridazin-3-yl)piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024962-500MG
101819-298EA 320 USD
101819-298
1-(6-Chloropyridazin-3-yl)piperidine-3-carboxylic acid
1-(6-Chloropyridazin-3-yl)piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O₂
MDL Number: MFCD07365330
CAS Number: 893755-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR