4-Bromo-5-methyl-1-phenylpyrazole-3-carboxylic acid

Supplier: Matrix Scientific

024961-250MG
101819-296EA 287.69 USD
101819-296
4-Bromo-5-methyl-1-phenylpyrazole-3-carboxylic acid
4-Bromo-5-methyl-1-phenylpyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉BrN₂O₂
MDL Number: MFCD09787497
CAS Number: 73227-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR