5-Bromo-2-fluoro-4-methylsulfonylaniline

Supplier: Matrix Scientific

024945-250MG
101819-260EA 306.15 USD
101819-260
5-Bromo-2-fluoro-4-methylsulfonylaniline
5-Bromo-2-fluoro-4-methylsulfonylaniline

Hazard: Irritant

Formula: C₇H₇BrFNO₂S
MDL Number: MFCD09842793
CAS Number: 1020722-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR