4-[(4-Methylphenyl)sulfonyl]benzaldehyde

Supplier: Matrix Scientific

024943-500MG
101819-256EA 364.62 USD
101819-256
4-[(4-Methylphenyl)sulfonyl]benzaldehyde
4-[(4-Methylphenyl)sulfonyl]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₂O₃S
MDL Number: MFCD09835170
CAS Number: 113823-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR