1-[(2-Fluoro-5-bromo-4-methylsulfonyl)phenyl]piperazine

Supplier: Matrix Scientific

024941-250MG
101819-252EA 306.15 USD
101819-252
1-[(2-Fluoro-5-bromo-4-methylsulfonyl)phenyl]piperazine
1-[(2-Fluoro-5-bromo-4-methylsulfonyl)phenyl]piperazine

Hazard: Irritant

Formula: C₁₁H₁₄BrFN₂O₂S
MDL Number: MFCD09839314
CAS Number: 1020722-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR