4,5-Dihydronaphtho[1,2-a]thiazol-2-ylamine

Supplier: Matrix Scientific

024937-500MG
101819-246EA 315.38 USD
101819-246
4,5-Dihydronaphtho[1,2-a]thiazol-2-ylamine
4,5-Dihydronaphtho[1,2-a]thiazol-2-ylamine

Hazard: Irritant

Formula: C₁₁H₁₀N₂S
MDL Number: MFCD02232183
CAS Number: 34176-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR