2-Amino-4-[(4-ethylsulfonyl)phenyl]-1,3-thiazole

Supplier: Matrix Scientific

024918-500MG
101819-212EA 401.54 USD
101819-212
2-Amino-4-[(4-ethylsulfonyl)phenyl]-1,3-thiazole
2-Amino-4-[(4-ethylsulfonyl)phenyl]-1,3-thiazole

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂S₂
MDL Number: MFCD02664035
CAS Number: 383129-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR