3-Fluoro-4-(methylsulfonyl)benzoic acid

Supplier: Matrix Scientific

024909-500MG
101819-194EA 404.62 USD
101819-194
3-Fluoro-4-(methylsulfonyl)benzoic acid
3-Fluoro-4-(methylsulphonyl)benzoic acid

Hazard: Irritant

Formula: C₈H₇FO₄S
MW: 218.2000 g/mol
MDL Number: MFCD09743730
CAS Number: 185945-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR