1-[(2-Methoxy-6-methylsulfonyl)phenyl]piperazine

Supplier: Matrix Scientific

024904-250MG
101819-184EA 315.38 USD
101819-184
1-[(2-Methoxy-6-methylsulfonyl)phenyl]piperazine
1-[(2-Methoxy-6-methylsulfonyl)phenyl]piperazine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃S
MDL Number: MFCD09750908
CAS Number: 1000018-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR