3-(3-Methylpiperidin-1-yl)-4-methylsulfonylaniline

Supplier: Matrix Scientific

024900-500MG
101819-176EA 476.92 USD
101819-176
3-(3-Methylpiperidin-1-yl)-4-methylsulfonylaniline
3-(3-Methylpiperidin-1-yl)-4-methylsulfonylaniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MDL Number: MFCD09743723
CAS Number: 1000018-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR